Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ABRO Rabbit anti-Human, Alexa Fluor 594, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Supplier:  Novus Biologicals NBP297533AF594

Catalog No. NB102551


Add to Cart

Description

Description

ABRO Polyclonal specifically detects ABRO in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

ABRO
Polyclonal
50 mM Sodium Borate
Rabbit
Antigen Affinity-purified
RUO
23172
Store at 4°C in the dark.
IgG
Western Blot, Immunofluorescence
Alexa Fluor 594
Abraxas brother protein 1, ABRO1abraxas brother 1, Em:AC068896.4, family with sequence similarity 175, member B, FLJ22338, KIAA0157BRISC complex subunit Abro1, Protein FAM175B
Produced in rabbits immunized with E. coli-derived Human ABRO fragment.
0.1 mL
Primary
Human
Purified
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit