Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(±)-Abscisic Acid 98%, ACROS Organics™

Click to view available options
Quantity:
100mg
500mg
Packaging:
Glass bottle
14375-45-2
C15H20O4
264.321
MFCD00075619
JLIDBLDQVAYHNE-LXGGSRJLSA-N
abscisic acid,+/--abscisic acid,acide abscissique,2z,4e-5-1-hydroxy-2,6,6-trimethyl-4-oxocyclohex-2-en-1-yl-3-methylpenta-2,4-dienoic acid,abscisinsaeure,abszisinsaeure,acido abscisico,+-abscisic acid,2-cis-abscisic acid,rs-abscisic acid
5375199
CHEBI:22152
(2Z,4E)-5-(1-hydroxy-2,6,6-trimethyl-4-oxocyclohex-2-en-1-yl)-3-methylpenta-2,4-dienoic acid
CC1=CC(=O)CC(C1(C=CC(=CC(=O)O)C)O)(C)C
14375-45-2
C15H20O4
264.321
MFCD00075619
JLIDBLDQVAYHNE-LXGGSRJLSA-N
abscisic acid,+/--abscisic acid,acide abscissique,2z,4e-5-1-hydroxy-2,6,6-trimethyl-4-oxocyclohex-2-en-1-yl-3-methylpenta-2,4-dienoic acid,abscisinsaeure,abszisinsaeure,acido abscisico,+-abscisic acid,2-cis-abscisic acid,rs-abscisic acid
5375199
CHEBI:22152
(2Z,4E)-5-(1-hydroxy-2,6,6-trimethyl-4-oxocyclohex-2-en-1-yl)-3-methylpenta-2,4-dienoic acid
CC1=CC(=O)CC(C1(C=CC(=CC(=O)O)C)O)(C)C
Sublimation Point 0120.0°C
Melting Point 188.0°C to 191.0°C
Quantity 100mg
Packaging Glass bottle
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Merck Index 15, 12
Formula Weight 264.32
Percent Purity 98%
Chemical Name or Material (±)-Abscisic acid, 98%
Show More Show Less

LIGHT SENSITIVE

TSCA : TSCA