Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abscisic Acid (Synthetic) 98.0+%, TCI America™

Click to view available options
Quantity:
100mg
500mg
14375-45-2
C15H20O4
264.321
MFCD00075619
JLIDBLDQVAYHNE-LXGGSRJLSA-N
abscisic acid,+/--abscisic acid,acide abscissique,2z,4e-5-1-hydroxy-2,6,6-trimethyl-4-oxocyclohex-2-en-1-yl-3-methylpenta-2,4-dienoic acid,abscisinsaeure,abszisinsaeure,acido abscisico,+-abscisic acid,2-cis-abscisic acid,rs-abscisic acid
5375199
CHEBI:22152
(2Z,4E)-5-(1-hydroxy-2,6,6-trimethyl-4-oxocyclohex-2-en-1-yl)-3-methylpenta-2,4-dienoic acid
CC1=CC(=O)CC(C1(C=CC(=CC(=O)O)C)O)(C)C
14375-45-2
C15H20O4
264.321
MFCD00075619
JLIDBLDQVAYHNE-LXGGSRJLSA-N
abscisic acid,+/--abscisic acid,acide abscissique,2z,4e-5-1-hydroxy-2,6,6-trimethyl-4-oxocyclohex-2-en-1-yl-3-methylpenta-2,4-dienoic acid,abscisinsaeure,abszisinsaeure,acido abscisico,+-abscisic acid,2-cis-abscisic acid,rs-abscisic acid
5375199
CHEBI:22152
(2Z,4E)-5-(1-hydroxy-2,6,6-trimethyl-4-oxocyclohex-2-en-1-yl)-3-methylpenta-2,4-dienoic acid
CC1=CC(=O)CC(C1(C=CC(=CC(=O)O)C)O)(C)C
Quantity 100mg
Melting Point 186°C
Color Undesignated
Formula Weight 264.32
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (HPLC,T)
Chemical Name or Material Abscisic Acid (Synthetic)

TSCA : No

Recommended Storage : Refrigerator