Novus Biologicals AbSelect TCS Purification Antibody Purification Kit

Fast and easy to use kit for purification of IgG fractions from hybridoma supernatants

Manufacturer: Novus Biologicals 8620500

 View more versions of this product

Catalog No. 86-205-00

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Specifications

    Product Type AbSelect TCS Purification Antibody Purification Kit
    Sufficient For 3 Purifications

    AbSelect TCS is prepared by coupling highly purified protein A to agarose beads which can be used to purify IgG fractions from hybridoma supernatants. The method involves the capture of the antibody on the AbSelect TCS resin and the removal of unwanted substances using a simple wash procedure. The purified product is then eluted and neutralized.

    Recommended Storage

    Store at 4°C. Do not freeze

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit