Learn More
Thermo Scientific™ ABsolute™ qPCR Master Mixes, SYBR Green-Based Detection
Thermo Scientific™ ABsolute qPCR Master Mixes offer sensitivity and reproducibility for real-time PCR applications.
Specifications
Storage Requirements -20°C Avoid freeze/thaw cycles
View More Specs
Products 2
Catalog Number Mfr. No. Description Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Description Quantity Price Quantity & Availability  
AB-1158A
View Documents
Thermo Scientific™
AB1158A
ABsolute QPCR SYBR Green Mix 100 x 50µL rxns This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
AB-1163B
View Documents
Thermo Scientific™
AB1163B
ABsolute QPCR SYBR Green ROX (500nM) Mix 2000 x 50µL rxns This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
Enjoy sensitivity and reproducibility in real-time PCR applications with Thermo Scientific™ ABsolute™ SYBR Green-Based Detection qPCR Master Mixes.
ABsolute qPCR Master Mixes are designed to achieve the most consistent and reproducible data across all qPCR platforms. Proprietary additives give enhanced and consistent end point readings as well as low Ct values. All mixes contain Thermo-Start Taq DNA Polymerase, for increased specificity, and contain dTTP for increased reaction efficiency and sensitivity (low Ct).
  • Maximum specificity—Chemically-modified Thermo-Start™ Taq remains completely inactive until heated to 95°C for 15 minutes, thereby preventing nonspecific amplification
  • Improved efficiency—The use of dTTP in place of dUTP increases reaction efficiency by approximately 5% and therefore reduces reaction time
  • Increased sensitivity—The combination of Thermo-Start Taq and dTTP increases sensitivity and allows for detection of low copy number templates
  • Instrument specific—Individually optimized qPCR mixes designed for all major qPCR / Real-Time PCR instruments
  • Reproducible results—All mixes undergo extensive quality control testing to ensure batch-to-batch consistency
  • Simple protocol—Master mix format reduces protocol steps and minimizes risk of pipetting errors and sample contamination
Includes:
ABsolute qPCR Buffer (Thermo-Start DNA polymerase, dNTPs, MgCl2, SYBR Green I, ROX™ passive reference dye (only in ROX mixes), Fluorescein (only in Fluorescein mix)); Separate MgCl2; Separate ROX vial (only in kits indicated)
Recommended for:
Ideal for QPCR analysis in molecular biology applications, including gene expression and SNP typing
Specifications
Specifications
-20°C Avoid freeze/thaw cycles
Purchase of this product includes an immunity from suit under patents specified in the product insert to use only the amount purchased for the purchaser's own internal research. No other patent rights are conveyed expressly, by implication, or by estoppel. Further information on purchasing licenses may be obtained by contacting the Director of Licensing, Applied Biosystems, 850 Lincoln Centre Drive, Foster City, California 94404, USA.