Tocris™ ABT 724 trihydrochloride

Potent, selective D4 partial agonist; proerectile

Manufacturer: Tocris™ 2214/50

 View more versions of this product

Catalog No. 22-145-0

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only


    Specifications

    Inhibitors ABT 724 trihydrochloride
    Purity >99
    Molecular Weight 402.75
    Quantity 50mg
    Product Type ABT 724 trihydrochloride

    Potent dopamine D4 receptor partial agonist (EC50 = 12.4 nM; 61% efficacy vs. dopamine). Has no agonist activity at D2 receptors (EC50 > 10µM). Selective in rats in vivo; produces penile erection following i.c.v. administration; increases intracavernosal pressure and potentiates the proerectile effects of sildenafil following s.c. administration. Displays minimal side effects.

    • Chemical Name: 2-[[4-(2-Pyridinyl)-1-piperazinyl]methyl]-1H-benzimidazole trihydrochloride
    • Formula: C17H19N5.3HCl
    • CAS Number: 587870-77-7
    • Solubility: Soluble to 100mM in water
    • Purity: >99

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit