ABT1 293T Cell Overexpression Lysate (Denatured), Abnova

Manufacturer: Abnova Corporation H00029777T02

 View more versions of this product

Catalog No. 89-002-148

  Check Availability
Add to Cart

    Specifications

    pH Range 6.8
    Plasmid pCMV-ABT1 full-length
    Quality Control Testing Transient overexpression cell Lysate was tested with Anti-ABT1 antibody (H00029777-B02) by western blotting
    Quantity 100μL
    Storage Buffer 1X Sample Buffer: 50mM Tris HCl, 2% SDS, 300mM 2-mercaptoethanol, 0.01% Bromophenol Blue, 10% Glycerol
    Storage Requirements -80°C
    Cell Line 293T
    For Use With (Application) WB
    Gene Accession No. NM_013375.2
    Host Species Human
    Molecular Weight 30.03

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit