Learn More
ABTB2 Rabbit anti-Human, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 243231AP
Catalog No. 50-173-4299
Add to cart
Description
Description
May be involved in the initiation of hepatocyte growth.
Specifications
Specifications
ABTB2
0.43 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q8N961
ABTB2
Rabbit
IgG
150 μL
-20°C
Primary
25841
Western Blot
Unconjugated
ABTB2
Liquid
ABTB2
ABTB2 Fusion Protein Ag19284
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human