Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ABTS 98.0+%, TCI America™

Click to view available options
Quantity:
1g
5g
30931-67-0
C18H24N6O6S4
548.666
MFCD00010404
OHDRQQURAXLVGJ-IXVYLBKPSA-N
ammonium 2,2'-hydrazine-1,2-diylidene bis 3-ethyl-2,3-dihydrobenzo d thiazole-6-sulfonate
91884754
diazanium;3-ethyl-2-[(E)-(3-ethyl-6-sulfonato-1,3-benzothiazol-2-ylidene)hydrazinylidene]-1,3-benzothiazole-6-sulfonate
CCN1C2=C(C=C(C=C2)S(=O)(=O)[O-])SC1=NN=C3N(C4=C(S3)C=C(C=C4)S(=O)(=O)[O-])CC.[NH4+].[NH4+]
30931-67-0
C18H24N6O6S4
548.666
MFCD00010404
OHDRQQURAXLVGJ-IXVYLBKPSA-N
ammonium 2,2'-hydrazine-1,2-diylidene bis 3-ethyl-2,3-dihydrobenzo d thiazole-6-sulfonate
91884754
diazanium;3-ethyl-2-[(E)-(3-ethyl-6-sulfonato-1,3-benzothiazol-2-ylidene)hydrazinylidene]-1,3-benzothiazole-6-sulfonate
CCN1C2=C(C=C(C=C2)S(=O)(=O)[O-])SC1=NN=C3N(C4=C(S3)C=C(C=C4)S(=O)(=O)[O-])CC.[NH4+].[NH4+]

[for Biochemical Research]

Color Green-White
Quantity 5g
Formula Weight 548.67
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (T)
Chemical Name or Material ABTS [=2,2′-Azinobis(3-ethylbenzothiazoline-6-sulfonic Acid Ammonium Salt)] [for Biochemical Research]

RTECSNumber : DL7002000

TSCA : No