Learn More
BioworldSupplier Diversity Partner ABUNDANT PROTEIN REMOVAL A DFS Item
Manufacturer:  BioworldSupplier Diversity Partner 220400001
ABUNDANT PROTEIN REMOVAL A-1-ANTITRYPSIN AB KIT: The bio-World Abundant Protein Removal a-1-antitrypsin kit removes the high-abundant protein a-1-antitrypsin from human serum and plasma. The high affinity-based column contains Separopor...
Catalog No. 50-252-999