Learn More
Bioworld Abundant Protein Removal-HSA Kit, 1 mL
Supplier:  Bioworld 220400021
Encompass_Preferred
The bio-World Abundant Protein Removal-HSA kit removes the high-abundant protein- HSA from human serum and plasma. The high affinity-based column contains Separopore-matrix coupled with antibodies against the serum protein. The presence of abundant proteins in human serum and plasma often masks the detection of low-abundant proteins of interest that serve as biomarkers for disease when being studied by 2D gel electrophoresis or LC/MS. After depletion of these high-abundant proteins, serum samples consist of an enriched pool of low-abundant proteins that can be further studied without significant interferences with abundant proteins. The Abundant Protein Removal-HSA kit provides a binding capacity of 2 mg abundant protein per column.Kit Components:• Anti-HSA Separopore, 1 ml column• Binding/Wash buffer, 25 mlRelated items:Catalog# 515909 removes serum haptoglobin onlyCatalog# 515908 removes serum transferrin onlyCatalog# 515907 removes alpha-1-antitrypsin
Catalog No. 50-253-001