ABX/Biochem Calibrators and Controls Available on GSA/VA Contract for Federal Government customers only.
Hematology analyzer controls and calibrators
Manufacturer: ABX Diagnostics, Inc. 5300000283
 View more versions of this product
Catalog No. 23-450-704
    N/A
This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance.  Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 
    Specifications
    Product Type Minotrol control; Micros 60
    For Use With (Application) Hematology controls
    For Use With (Equipment) ABX Micros SIM2
    Quantity 12 x 2.5mL
    Level 3
  • Description & Specifications
Related Products
Product Content Feedback