Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AC 45594, Tocris Bioscience™

Click to view available options
Quantity:
10mg
50mg
This item is not returnable. View return policy
13037-86-0
C13H20O2
208.301
HZBABTUFXQLADL-UHFFFAOYSA-N
4-heptyloxyphenol,4-heptyloxy phenol,p-heptyloxy phenol,phenol, 4-heptyloxy,p-n-heptyloxyphenol,4-n-heptyloxyphenol,p-heptyloxyphenol,4-heptyloxy-phenol,unii-w9gse02182,4-heptyloxy benzolol
25641
CHEBI:34406
4-heptoxyphenol
CCCCCCCOC1=CC=C(C=C1)O
This item is not returnable. View return policy
13037-86-0
C13H20O2
208.301
HZBABTUFXQLADL-UHFFFAOYSA-N
4-heptyloxyphenol,4-heptyloxy phenol,p-heptyloxy phenol,phenol, 4-heptyloxy,p-n-heptyloxyphenol,4-n-heptyloxyphenol,p-heptyloxyphenol,4-heptyloxy-phenol,unii-w9gse02182,4-heptyloxy benzolol
25641
CHEBI:34406
4-heptoxyphenol
CCCCCCCOC1=CC=C(C=C1)O

SF-1 inverse agonist

Selective inverse agonist at the orphan nuclear receptor steroidogenic factor-1 (SF-1) (IC 50 = 50 - 100 nM). Displays no activity at estrogen, LRH-1, ROR, ERR or Nurr receptors. Inhibits SFRE-mediated transcription.
Quantity 50mg
Formula Weight 208.3
Percent Purity >99%
Chemical Name or Material AC 45594

Recommended Storage : Store at +4°C