Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AC 55649, Tocris Bioscience™

Click to view available options
Quantity:
10mg
50mg
This item is not returnable. View return policy
59662-49-6
C21H26O2
310.437
HXBKPYIEQLLNBK-UHFFFAOYSA-N
4-4-octylphenyl benzoic acid,4'-octyl-4-biphenylcarboxylic acid,4-octyl-4'-biphenylcarboxylic acid,8bca,4'-octyl-biphenyl-4-carboxylic acid,4'-n-octylbiphenyl-4-carboxylic acid,4-n-octylbiphenyl-4'-carboxylic acid,4'-octyl-1,1'-biphenyl-4-carboxylic acid,4'-octylbiphenyl-4-carboxylic acid,1,1'-biphenyl-4-carboxylicacid, 4'-octyl
1714884
4-(4-octylphenyl)benzoic acid
CCCCCCCCC1=CC=C(C=C1)C2=CC=C(C=C2)C(=O)O
This item is not returnable. View return policy
59662-49-6
C21H26O2
310.437
HXBKPYIEQLLNBK-UHFFFAOYSA-N
4-4-octylphenyl benzoic acid,4'-octyl-4-biphenylcarboxylic acid,4-octyl-4'-biphenylcarboxylic acid,8bca,4'-octyl-biphenyl-4-carboxylic acid,4'-n-octylbiphenyl-4-carboxylic acid,4-n-octylbiphenyl-4'-carboxylic acid,4'-octyl-1,1'-biphenyl-4-carboxylic acid,4'-octylbiphenyl-4-carboxylic acid,1,1'-biphenyl-4-carboxylicacid, 4'-octyl
1714884
4-(4-octylphenyl)benzoic acid
CCCCCCCCC1=CC=C(C=C1)C2=CC=C(C=C2)C(=O)O

Selective RARβ2 agonist

Potent, isoform-selective RARβ2 receptor agonist (pEC 50 values are 6.9, 5.7 and 5.6 at RARβ2, RARβ1 and RARα respectively) that displays 100-fold selectivity versus other retinoid receptors. Inhibits proliferation of the breast cancer cell line MCF-7.
Quantity 10mg
Formula Weight 310.44
Percent Purity >99%
Chemical Name or Material AC 55649

Recommended Storage : Store at +4°C