Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(±)-AC 7954 hydrochloride, Tocris

Non-peptide UT receptor agonist

Manufacturer:  R&D Systems 2484/10

Quantity 10mg

 View more versions of this product

Catalog No. 24-841-0


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

>98%
366.28
(±)-AC 7954 hydrochloride
C19H20ClNO2·HCl
(±)-3-(4-Chlorophenyl)-3-[2-(dimethylamino)ethyl]-3, 4-dihydro-1H-2-benzopyran-1-one hydrochloride

Specifications

10mg
>98%
366.28
(±)-AC 7954 hydrochloride
477313-09-0
C19H20ClNO2·HCl
(±)-3-(4-Chlorophenyl)-3-[2-(dimethylamino)ethyl]-3, 4-dihydro-1H-2-benzopyran-1-one hydrochloride

Safety and Handling

Recommended Storage : Store at +4°C

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit