Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Ac-LEU-LEU-TYR-7-Amino-4-Trifluoromethylcoumarin

A fluorogenic substrate for Calpain

$639.50 - $1450.00

Specifications

Format Solid
Color White
Molecular Weight 660
For Use With (Application) Cell Migration Analysis, Cell Signaling Analysis
Storage Requirements Keep cool and dry
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MP3AFC11610
View Documents
MP Biomedicals™
03AFC11610
10mg
Each for $639.50
Add to cart
 
MP3AFC11625
View Documents
MP Biomedicals™
03AFC11625
25mg
Each for $1,450.00
Add to cart
 
Description

Description

The AFC substrate is both chromogenic and fluorogenic. The specificity and sensitivity of AFC allows for the detection of enzyme activities in cell lysate and electrophoretic gels as the emission spectra of free and the conjugated AFC do not overlap. Excitation 400nm; Emission 505nm

Specifications

Specifications

Solid
660
Keep cool and dry
White
Cell Migration Analysis, Cell Signaling Analysis
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit