Learn More
Tocris Bioscience™ Ac2-12
Annexin/lipocortin 1-mimetic; inhibits leukocyte extravasation
Manufacturer:  Tocris Bioscience™ 1846/1
Catalog No. 18-461-R
Add to cart
This item is not returnable. View return policy
Description
Description
Annexin/lipocortin 1-mimetic peptide; inhibits leukocyte extravasation. Reduces neutrophil adhesion and emigration, and promotes detachment of neutrophils from activated mesenteric endothelium in mice in vivo.
Specifications
Specifications
Ac2-12
1mg
Ac2-12
1351.58
Ac-Ala-Met-Val-Ser-Glu-Phe-Leu-Lys-Gln-Ala-Trp