Learn More
Abnova™ ACAA1 293T Cell Transient Overexpression Lysate (Denatured) (T01)
Human overexpression lysate
Manufacturer:  Abnova™ H00000030T01
Catalog No. 89-932-259
Add to cart
Description
Description
Specifications
Specifications
293T
NM_001607.2
pCMV-ACAA1 full-length
100 μL
Human
ACAA1 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 46.75kDa
Transient overexpression cell lysate was tested with anti-ACAA1 antibody (Mfr. No. H00000030-B01) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Western Blot