Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ACAA2 Antibody (OTI1C10), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP246477
Description
Ensure accurate, reproducible results in Western Blot, Immunohistochemistry, Immunofluorescence
ACAA2 Monoclonal specifically detects ACAA2 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence.Specifications
| ACAA2 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:200 - 1:4000, Immunohistochemistry 1:150, Immunofluorescence 1:100 | |
| 3-ketoacyl-CoA thiolase, mitochondrial, Acetyl-CoA acyltransferase, acetyl-CoA acyltransferase 2, acetyl-Coenzyme A acyltransferase 2, beta ketothiolase, beta-ketothiolase, DSAEC, EC 2.3.1, EC 2.3.1.16, FLJ35992, FLJ95265, Mitochondrial 3-oxoacyl-CoA thiolase, mitochondrial 3-oxoacyl-Coenzyme A thiolase, T1 | |
| Mouse | |
| 41.7 kDa | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat, Canine, Monkey | |
| Purified |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| OTI1C10 | |
| Unconjugated | |
| PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide | |
| ACAA2 | |
| Full length human recombinant protein of human ACAA2(NP_006102) produced in HEK293T cell. | |
| Immunogen affinity purified | |
| RUO | |
| 10449 | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction