Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acacia, Milled Powder, NF, Spectrum™

Click to view available options
Quantity:
100 g
500 g
2.5 kg
12 kg
45 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Bottle
Poly Pail
9000-01-5
9000-01-5

GU117, 9000-01-5

Spectrum™ Acacia, Milled Powder, NF is a natural gum used primarily in the food industry as a stabilizer. It is a mixture of polysaccharides and glycoproteins giving it the properties of a glue and binder which is edible by humans. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Poly Bottle
Quantity 500 g
CAS Max % 100%