Learn More
ACAD9 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 157701AP
Catalog No. 50-173-0464
Add to cart
Description
Description
This gene encodes a member of the acyl-CoA dehydrogenase family. Members of this family of proteins localize to the mitochondria and catalyze the rate-limiting step in the beta-oxidation of fatty acyl-CoA. The encoded protein is specifically active toward palmitoyl-CoA and long-chain unsaturated substrates. Mutations in this gene cause acyl-CoA dehydrogenase family member type 9 deficiency. Alternate splicing results in multiple transcript variants.
Specifications
Specifications
ACAD9
0.31 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8JZN5, Q9H845
ACAD 9, ACAD9
Rabbit
IgG
150 μL
-20°C
Primary
229211, 28976, 294973
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
ACAD9
Liquid
ACAD9
ACAD9 Fusion Protein Ag8414
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat