Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACADM Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP15472920UL

 View more versions of this product

Catalog No. NBP15472920


Add to cart

Description

Description

ACADM Polyclonal antibody specifically detects Antigen in Human samples. It is validated for Western Blotting.
Specifications

Specifications

ACADM
Unconjugated
PBS & 2% Sucrose. with No Preservative
Affinity Purified
ACADM
Synthetic peptides corresponding to ACADM(acyl-Coenzyme A dehydrogenase, C-4 to C-12 straight chain) The peptide sequence was selected from the N terminal of ACADM. Peptide sequence AAGFGRCCRVLRSISRFHWRSQHTKANRQREPGLGFSFEFTEQQKEFQAT.
46 kDa
20ul
Lipid and Metabolism
Primary
34
Western Blot
Western Blot 0.2-1 ug/ml
P11310
ACAD1, acyl-CoA dehydrogenase, C-4 to C-12 straight chain, acyl-Coenzyme A dehydrogenase, C-4 to C-12 straight chain, EC 1.3.99, EC 1.3.99.3, FLJ18227, FLJ99884, MCADFLJ93013, MCADH, medium-chain specific acyl-CoA dehydrogenase, mitochondrial
Rabbit
IgG
Immunogen affinity purified
RUO
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit