Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACADM Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$152.22 - $378.35

Specifications

Antigen ACADM
Immunogen Synthetic peptides corresponding to ACADM(acyl-Coenzyme A dehydrogenase, C-4 to C-12 straight chain) The peptide sequence was selected from the N terminal of ACADM. Peptide sequence AAGFGRCCRVLRSISRFHWRSQHTKANRQREPGLGFSFEFTEQQKEFQAT.
Host Species Rabbit
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP15472920
View Documents
Novus Biologicals
NBP15472920UL
20ul Each for $152.22
Add to cart
 
NBP154729
View Documents
Novus Biologicals
NBP154729
100 ul Each for $378.35
Add to cart
 
Description

Description

ACADM Polyclonal antibody specifically detects Antigen in Human samples. It is validated for Western Blotting.
Specifications

Specifications

ACADM
Rabbit
Polyclonal
Unconjugated
34
Affinity Purified
Western Blot
RUO
Synthetic peptides corresponding to ACADM(acyl-Coenzyme A dehydrogenase, C-4 to C-12 straight chain) The peptide sequence was selected from the N terminal of ACADM. Peptide sequence AAGFGRCCRVLRSISRFHWRSQHTKANRQREPGLGFSFEFTEQQKEFQAT.
Primary
IgG
ACAD1, acyl-CoA dehydrogenase, C-4 to C-12 straight chain, acyl-Coenzyme A dehydrogenase, C-4 to C-12 straight chain, EC 1.3.99, EC 1.3.99.3, FLJ18227, FLJ99884, MCADFLJ93013, MCADH, medium-chain specific acyl-CoA dehydrogenase, mitochondrial
PBS & 2% Sucrose. with 0.09% Sodium Azide
Immunogen affinity purified
Store at -20C. Avoid freeze-thaw cycles.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit