Learn More
Aviva Systems Biology Corp ACADS Antibody - N-terminal region (ARP54275_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP54275_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ACADS. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ACADS. Alternate names: ACAD3, SCADApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-61155