Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ACADS Mouse anti-Human, Clone: 8B10, Novus Biologicals™
Mouse Monoclonal Antibody
$337.00 - $482.50
Specifications
Antigen | ACADS |
---|---|
Clone | 8B10 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:1000, ELISA, Immunocytochemistry/Immunofluorescence 1:100 |
Applications | Western Blot, ELISA, Immunocytochemistry, Immunofluorescence |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP25949650
|
Novus Biologicals
NBP25949650UL |
50 μL |
Each for $337.00
|
|
|||||
NBP259496X
|
Novus Biologicals
NBP259496100UL |
100 μL |
Each for $482.50
|
|
|||||
Description
ACADS Monoclonal specifically detects ACADS in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
ACADS | |
1 mg/mL | |
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
Mouse | |
Human | |
ACAD3, acyl-CoA dehydrogenase, C-2 to C-3 short chain, acyl-Coenzyme A dehydrogenase, C-2 to C-3 short chain, Butyryl-CoA dehydrogenase, EC 1.3.99, EC 1.3.99.2, mitochondrial short-chain specific acyl-CoA dehydrogenase, SCADshort-chain acyl-CoA dehydrogenase, short-chain specific acyl-CoA dehydrogenase, mitochondrial, unsaturated acyl-CoA reductase | |
ACADS | |
IgG2a κ | |
Protein A purified |
8B10 | |
Western Blot 1:1000, ELISA, Immunocytochemistry/Immunofluorescence 1:100 | |
Monoclonal | |
Purified | |
RUO | |
Liquid. In PBS (pH 7.4), 10% Glycerol with 0.02% Sodium Azide | |
35 | |
Recombinant human ACADS (25-412aa) purified from E. coli | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title