Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACADS Mouse anti-Human, Non-human primate, Clone: OTI1D2, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA526440

Catalog No. PIMA526440


Add to cart

Description

Description

Introduces a double bond at position 2 in saturated acyl-CoA ft.s of short chain length, i.e. less than 6 carbon atoms.
Specifications

Specifications

ACADS
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
P16219
ACAD3, SCAD
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
35, 698196
Immunohistochemistry (Paraffin), Western Blot
OTI1D2
Unconjugated
ACADS
Liquid
ACADS
Human recombit protein fragment corresponding to amino acids 34-260 of ACADS produced in E.coli
Affinity Chromatography
RUO
Antibody
Monoclonal
Human, Non-human primate
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit