Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACAP3 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$436.00

Specifications

Antigen ACAP3
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP170400
View Documents
Novus Biologicals
NBP170400
100 μL
Each of 1 for $436.00
Add to cart
 
Description

Description

ACAP3 Polyclonal specifically detects ACAP3 in Human samples. It is validated for Western Blot.
Specifications

Specifications

ACAP3
Polyclonal
Rabbit
PBS and 2% Sucrose with 0.09% Sodium Azide
116983
Synthetic peptides corresponding to CENTB5(centaurin, beta 5) The peptide sequence was selected from the N terminal of CENTB5. Peptide sequence LQSFVKEDVRKFKETKKQFDKVREDLELSLVRNAQAPRHRPHEVEEATGA.
Primary
Store at -20C. Avoid freeze-thaw cycles.
Western Blot
Unconjugated
RUO
ANK repeat and PH domain-containing protein 3, ArfGAP with coiled-coil, ankyrin repeat and PH domains 3, centaurin, beta 5, centaurin-beta-5, CENTB5, cnt-b5, KIAA1716
ACAP3
IgG
Affinity Purified
85 kDa
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit