Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACAT1 Mouse anti-Human, Alexa Fluor™ 532, Clone: 1500000, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP222565AF532

Catalog No. NP2226A32


Add to cart

Description

Description

ACAT1 Monoclonal antibody specifically detects ACAT1 in Human samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

ACAT1
1500000
Western Blot, ELISA
Conjugated, Purified
ACAT1
Recombinant human ACAT1 (34-427aa) purified from E. coli
Protein A purified
Cancer, Cellular Markers, Lipid and Metabolism, Mitochondrial Markers, Signal Transduction
Primary
38.0
ELISA, Western Blot
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
ACAT, acetoacetyl Coenzyme A thiolase, Acetoacetyl-CoA thiolase, acetyl-CoA acetyltransferase 1, acetyl-CoA acetyltransferase, mitochondrial, acetyl-Coenzyme A acetyltransferase 1, EC 2.3.1, EC 2.3.1.9, MAT, mitochondrial acetoacetyl-CoA thiolase, T2, THIL
Mouse
IgG1 Kappa
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit