Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACAT2 Mouse anti-Human, Rat, Canine, Monkey, PE-Cyanine7, Clone: OTI3E2, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270068PECY7

Catalog No. N27068PECY7


Add to cart

Description

Description

ACAT2 Monoclonal antibody specifically detects ACAT2 in Human, Rat, Canine, Monkey samples. It is validated for Flow Cytometry.
Specifications

Specifications

ACAT2
OTI3E2
Flow Cytometry
Affinity Purified, Conjugated
ACAT2
Full length recombinant protein of human ACAT2 (NP_005882) produced in HEK293T cells.
Immunogen affinity purified
Cholesterol Metabolism, Lipid and Metabolism
Primary
39.0
Flow Cytometry
PE-Cyanine7
PBS with 0.05% Sodium Azide
acetoacetyl Coenzyme A thiolase, acetyl-CoA acetyltransferase 2, acetyl-CoA acetyltransferase, cytosolic, Acetyl-CoA transferase-like protein, acetyl-Coenzyme A acetyltransferase 2, acetyl-Coenzyme A acetyltransferase 2 (acetoacetyl Coenzyme A thiolase), ACTL, Cytosolic acetoacetyl-CoA thiolase, EC 2.3.1, EC 2.3.1.9
Mouse
IgG2b
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Canine, Human, Monkey, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit