Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACAT2 Mouse anti-Canine, Human, Rat, Clone: OTI7B1, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA525114

Catalog No. PIMA525114


Add to cart

Description

Description

Acetyl-Coenzyme A acetyltransferase 2 is an enzyme involved in lipid metabolism. Reported patients with ACAT2 deficiency have shown severe mental retardation and hypotonus. The ACAT2 gene shows complementary overlapping with the 3-prime region of the TCP1 gene in both mouse and human. These genes are encoded on opposite strands of DNA, as well as in opposite transcriptional orientation.
Specifications

Specifications

ACAT2
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q5XI22, Q9BWD1
Acetyl-CoA acetyltransferase, cytosolic; Cytosolic acetoacetyl-CoA thiolase; Acetyl-CoA transferase-like protein; ACTL; ACAT2
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
308100, 39, 484063
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
OTI7B1
Unconjugated
ACAT2
Liquid
ACAT2
Full length human recombit protein of ACAT2 produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Canine, Human, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit