Learn More
ACBD3 Mouse anti-Canine, Human, Mouse, Non-human primate, Rat, Clone: OTI1G2, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA525989
Catalog No. PIMA525989
Add to cart
Description
Description
GOLPH1, also known as GCP60, was initially identified as a Golgi protein that can interact with the integral membrane protein giantin and is thought to be involved in the maintece of the Golgi structure. GOLPH1 has also been shown to interact with other Golgi proteins such as Golgin-160, a Golgi protein that can be cleaved by caspases-2, -3, and -7, leading to the nuclear localization of Golgin-160. GOLPH1 interaction with the Golgin-160 fragments is stronger than that with the intact Golgin-160, with its interaction regulated by the oxidation state of Cys-463 within GOLPH1, suggesting that the nuclear localization of the caspase-cleaved Golgin-160 fragments is a highly coordinated event. GOLPH1 has also been found to interact with Numb, a cytosolic signaling protein that mediates asymetric cell division of neural progenitor cells to a daughter progenitor cell and a neuron, suggesting that Golgi fragmentation and reconstitution during the cell cycle differentially regulate Numb signaling through changes in GOLPH1 subcellular distribution and may couple cell fate with cell cycle progression.
Specifications
Specifications
ACBD3
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q9H3P7, Q8BMP6, Q7TNY6
GCP60, GOCAP1, GOLPH1, PAP7; Golgi resident protein GCP60; PBR- and PKA-associated protein 7; PKA (RIalpha)-associated protein; acyl-Coenzyme A binding domain containing 3; golgi complex associated protein 1, 60kDa; golgi phosphoprotein 1; peripheral benzodiazepine receptor-associated protein PAP7
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
170760, 289312, 611888, 64746, 699311
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
OTI1G2
Unconjugated
ACBD3
Liquid
ACBD3
Full length human recombit protein of ACBD3 produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Canine, Human, Mouse, Non-human primate, Rat