Learn More
Aviva Systems Biology Corp ACBD4 Antibody - N-terminal region (ARP49858_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP49858_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ACBD4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ACBD4. Alternate names: FLJ13322, FLJ90623, HMFT0700ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-60201