Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ACBD4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA551189
Description
The antibody detects endogenous levels of total ACBD4 protein.
ACBD4 is a member of the acyl-coenzyme A binding domain containing protein family. All family members contain the conserved acyl-Coenzyme A binding domain, which binds acyl-CoA thiol esters. They are thought to play roles in acyl-CoA dependent lipid metabolism.
Specifications
ACBD4 | |
Polyclonal | |
Unconjugated | |
ACBD4 | |
2010009P05Rik; 2010015A21Rik; Acbd4; acyl-CoA binding domain containing 4; acyl-CoA-binding domain-containing protein 4; acyl-Coenzyme A binding domain containing 4; AI849317; FLJ13322; FLJ90623; HMFT0700; similar to acyl-Coenzyme A binding domain containing 4; zgc:85611 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79777 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q8NC06 | |
ACBD4 | |
Full length fusion protein of human ACBD4. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction