Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACBD7 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548710

Catalog No. PA548710

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Binds medium- and long-chain acyl-CoA esters.
Specifications

Specifications

ACBD7
Polyclonal
PBS with 0.09% sodium azide
Q8N6N7
Acyl-CoA-binding domain-containing protein 7, ACBD7
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
414149
Western Blot
Unconjugated
ACBD7
Liquid
ACBD7
KLH conjugated synthetic peptide between 60-88 amino acids from the C-terminal region of human ACBD7.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit