Learn More
GSC Go Science Crazy Acceleration Motion Kit Products for Science Education
Complete Acceleration Kit with Photogates
Manufacturer:  GSC Go Science Crazy ACCMT
Catalog No. S24652
Add to cart
Description
Description
The Acceleration Motion Kit is used to study the motion of planes and slopes. The kit consists of a 1 meter long horizontal aluminum track with a graduated 0mm to 900mm scale, three adjustable supports for the track, two photogates, and a level adjuster knob that is used to increase the slope. Includes: • 1 Track • 1 Solenoid, 5V • 1 Solid steel physics balls, 19mm (#42036) • 2 Photogates
Specifications
Specifications
Education
TMRSOl