Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Axygen™ Accessories for Axygen™ Horizontal Gel Box, 10cm

For use with horizontal gel box, 10cm

Manufacturer:  Axygen™ HGB2020MC075

 View more versions of this product

Catalog No. 14-223-953


Add to cart

Specifications

Specifications

0.75mm
20-well Comb, 0.75mm thickness
Axygen Horizontal Gel Box, 20cm
20
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit