Learn More
Fisherbrand™ Accessories for Tube Rotator
14- to 63-place carousels for 11 to 34mm tubes
$215.00
Specifications
For Use With (Equipment) Fisher Scientific tube rotator
Product Type Carousel
View More Specs
Products 5
Catalog Number Diameter (Metric) Price Quantity & Availability  
Catalog Number Diameter (Metric) Price Quantity & Availability  
05-450-201
View Documents
27 to 34mm
Each for $215.00
Add to cart
05-450-204
View Documents
14mm
Each for $215.00
Add to cart
05-450-202
View Documents
9 to 11mm
Each for $215.00
Add to cart
05-450-203
View Documents
14 to 18mm
Each for $215.00
Add to cart
05-450-205
View Documents
18.5mm
Each for $215.00
Add to cart
Description
Description
Tube Rotator is Ideal for Biochemistry, Molecular Biology and Clinical applications. 14- to 63-place carousels for 11 to 34mm tubes
  • Tube carousel
Specifications
Specifications
Fisher Scientific tube rotator
Carousel