Learn More
Thermo Scientific™ Accessories for MaxQ™ 7000 Water Bath Shakers
Optimize the Accessories for Thermo Scientific™ MaxQ™ 7000 Water Bath Shakers to increase the versatility of the MaxQ 7000.
$388.00 - $1535.00
Products 4
Catalog Number Mfr. No. Description Price Quantity & Availability  
Catalog Number Mfr. No. Description Price Quantity & Availability  
11-675-462
View Documents
Thermo Scientific™
SHK700032
Plexiglas™ gable cover for temperatures up to 65°C
Each for $1,313.00
Add to Cart
11-675-464
View Documents
Thermo Scientific™
SHK700037
Cooling coil for near-ambient temperature and below (requires a circulating chiller)
Each for $410.50
Add to Cart
11-675-465
View Documents
Thermo Scientific™
SHK700040
Auxiliary platform suspension arms for rapid platform changes
Each for $388.00
Add to Cart
11-675-466
View Documents
Thermo Scientific™
SHK700045
Movable dolly for underLab bench operation
Each for $1,535.00
Add to Cart
Description
Description
Put Thermo Scientific™ Accessories for MaxQ™ 7000 Water Bath Shakers to use to increase the utility of MaxQ 7000 Water Bath Shakers. With exceptional innovation, reliability and accuracy, Thermo Scientific MaxQ 7000 Shakers, Platforms and accessories are the optimal choice for your busy lab.