Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ Replacement Chart Recorder Paper
For Use With: MaxQ™ Single Pen Chart Recorders
Supplier: Thermo Scientific™ 180006
Description
Integrate Thermo Scientific™ Data Output Accessories for your environmental chamber or incubator to increase functionality and compatibility.
Specifications
Chart paper for single pen chart recorders | |
LED | |
Accessory, Replacement Chart Recorder Paper |
MaxQ™ Single Pen Chart Recorders | |
0°C to 60°C | |
Customer Installed |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction