Learn More
Thermo Scientific™ Accessories for MaxQ™ HP Orbital Shakers Available on GSA/VA Contract for Federal Government customers only.
Get maximum flexibility, regardless of your application, by using these accessories for Thermo Scientific™ MaxQ™ High Performance orbital shakers.
Manufacturer:  Thermo Scientific™ 452002
 View more versions of this product
Catalog No. 452002
Add to cart
Description
Description
Get maximum flexibility, regardless of your application, by using Thermo Scientific™ Accessories for MaxQ™ High Performance Orbital Shakers.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Kneewell slide mount, shaker slides out on rails for easy access
83.8cm
56.4cm
MaxQ HP 420
MaxQ HP 420
33 in.
37.3 in.
Kneewell slide mount
Slide Mount