Learn More
MilliporeSigma™ Accessories to Milli-Q™ IQ 7000 Ultrapure Water System
Our Lab Water purification system portfolio offers a broad range of pure and ultrapure water purification systems and services designed for scientists working in pharmaceutical, clinical, academic, industrial, research, and government laboratories -- in both validated and non-validated environments.
$331.00 - $656.00
Specifications
For Use With (Equipment) Milli-Q IQ 7000 Ultrapure Water System
View More Specs
Products 3
Catalog Number Mfr. No. Type Price Quantity & Availability  
Catalog Number Mfr. No. Type Price Quantity & Availability  
ZFC0NDCA1
View Documents
MilliporeSigma™
ZFC0NDCA1
Feed Water Conductivity Cell
Each for $656.00
Add to Cart
ZMQSFTSA1
View Documents
MilliporeSigma™
ZMQSFTSA1
Foot Pedal
Each for $390.00
Add to Cart
ZWATSENA1
View Documents
MilliporeSigma™
ZWATSENA1
Water Sensor
Each for $331.00
Add to Cart
Description
Description
Water purification accessories.
Feed Water Conductivity Cell: Measures the conductivity of water feeding
Foot Pedal: Hands-free dispensing accessory
Tank Level Adapter:  For pure feed water stored in lab water storage tank 
Water Sensor: Detects water spillage and automatically closes the Milli-Q™ inlet solenoid valve to protect the surroundings from water damage; 120V/60Hz
Order Info
Must be ordered with the Milli-Q™ IQ 7000 Ultrapure Water System. Please contact your Fisher Rep for more details.
Specifications
Specifications
Milli-Q IQ 7000 Ultrapure Water System