Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OHAUS™ Accessories for Valor 3000 Xtreme Portable Food Portioning Scales

For use with Valor 3000 Xtreme Portable Food Portioning Scales.

Manufacturer:  OHAUS™ 80251216

 View more versions of this product

Catalog No. 01-919-400


Add to cart

Description

Description

Sandwich Pan
  • Stainless steel
  • For weighing sandwiches and other long food items
  • Clips over the platform of the Valor 3000 Xtreme
  • Measures 11W x 5.9D x 0.4H in. (28 x 15 x 1cm)
French Fry Holder
  • Stainless steel
  • For weighing french fries, onion rings, tacos, and other food service items that are required to remain upright during weighing
  • Clips over the platform of the Valor 3000 Xtreme
  • Dimensions, front wall: 4.75 in. (12.1cm) wide by 1.13 in. (2.9cm) high; back wall: 4.75 in. (12.1cm) wide by 2.75 in. (7cm) high
Ice Cream Cone Holder
  • Stainless steel
  • Used for weighing ice cream, frozen custard, gelati, and other food service items where the cone has to be held to keep the item upright
  • Clips over the platform of the Valor 3000 Xtreme
  • Upper platform measures 5.9 in. (15cm) wide by 3.5 in. (8.9cm) deep
  • There are two holes in the upper platform, one 2.25 in. (5.7cm) and the other 1.5 in. (3.8cm)
  • Height of upper platform is 4 in. (10.2cm) above lower platform
Specifications

Specifications

Carrying Case
Valor™ 3000 Xtreme digital compact scale
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit