Learn More
Accessories and Replacement Parts for Sartorius™ Balances: Switches Available on GSA/VA Contract for Federal Government customers only.
For remote control
Manufacturer:  Sartorius YFS01
 View more versions of this product
Catalog No. 14-557-533
  Check Availability
Add to cart
Specifications
Description
Hand and foot switch accessories for Sartorius balances.
Description & Specifications
Specifications
Foot Switch
CPA balance, extended balance, Master pro LA balance, SE and ME ultramicro and microbalance, M-power balance
T-connector