Learn More
Thermo Scientific™ Accessories for the Barnstead™ TII Type 2 Water System
Integrate the accessories for the Thermo Scientific™ Barnstear™ TII Type 2 Water System with the system itself for a more complete laboratory solution.
$1265.00
Specifications
Depth (English) 18.5 in.
Depth (Metric) 47cm
Bacterial Content <1CFU/mL
Resistivity 15MΩ·cm at 25°C
TOC Monitor with UV Intensity Monitoring No
View More Specs
Products 1
Catalog Number Mfr. No. Description Price Quantity & Availability  
Catalog Number Mfr. No. Description Price Quantity & Availability  
09-036-21
View Documents
Thermo Scientific™
AY1406X2
Smart Distribution Pump Each for $1,265.00
Add to cart
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Description
Description
Integrate the accessories for the Thermo Scientific™ Barnstear™ TII Type 2 Water System such as distribution pumps and UV lamps with the system itself to create a more complete lab solution.
Specifications
Specifications
18.5 in.
<1CFU/mL
No
100/240VAC 50/60Hz
19.5 in.
50/60Hz
1L/min.
100/240VAC
34.3cm
47cm
15MΩ·cm at 25°C
1 to 5ppb
30 to 100psig (2 to 6.9 bar)
49.5cm
4° to 40°C
12L/hr.
13.5 in.