Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medica Accessories for the EasyStat Blood Gas Analyzer

For use with the EasyStat Blood Gas Analyzer (Cat. No. 22-515-457, shown at left).

Manufacturer:  Medica 7201

 View more versions of this product

Catalog No. 22-515-459

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

A wide range of accessories for use with EasyStat Blood Gas Analyzer.
Specifications

Specifications

pH Electrode
EasyStat Blood Gas Analyzer
Each
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit