Learn More
MilliporeSigma™ Accessories for YY3009000 Filter Holder, 90 mm
Accessories for filter holders for sample preparation.
Manufacturer:  MilliporeSigma™ YY2214257
 View more versions of this product
Catalog No. YY2214257
Add to cart
Description
Description
Accessories for filter holders for sample preparation.
AccessoriesXX4204708 Vent/Relief ValveYY3009053 O-rings, 90 mm filter housing, PTFEYY3009054 Support Screen, PTFE-coatedYY2209058 Underdrain Support, 90 mmYY2209059 Support Legs
Specifications
Specifications
90 mm filter holders
Non-sterile
Hand Knob
Glass Filled PP, Brass
Hand Knob
Not Autoclavable