Learn More
IKA™ Accessory Stirring Shafts for Fisher Scientific™ SteadyStir™ and Overhead Stirrers
Generate axial flow in the vessel with these stirring shafts. IKA™ Accessory Stirring Shafts for Fisher Scientific™ SteadyStir™ and IKA™ Overhead Stirrers are designed for medium to high speeds.
Manufacturer:  IKA™ 2332900
 View more versions of this product
Catalog No. 14-259-069
Add to cart
Description
Description
Four-Bladed Propeller Stirrers
  • Standard stirring element
  • For drawing material to be mixed from top to bottom
  • Local shearing force
  • Generates axial flow
  • Use at medium to high speeds
Three-Bladed Propeller Stirrers
  • Flow efficient design
  • For drawing material to be mixed from top to bottom of vessel
  • Minimum shearing
  • Medium to high speeds
Turbine Stirrers
  • For drawing material to be mixed from above
  • Axial flow
  • Minimal danger of injury
  • Medium to high speeds
Dissolver Stirrer
  • Radial flow for drawing material to be stirred from top and bottom of vessel
  • High turbulance, high shearing forces
  • For particle reduction
  • Minimum and high speeds
Centrifugal Stirrers: Two bladed
  • Blades open with increasing speed
  • For stirring in round vessels with narrow necks
  • Medium to high speeds
Paddle Stirrers
  • Tangential flow, minimal turbulance, good heat exchange, gentle treatment of product
  • Low to medium speeds
Anchor Stirrers
  • Ideal stirrers for medium-to-high-viscosity fluids
  • Tangential flow, high shearing rate at edges, minimal deposits
  • Low speeds
Key to IKA models in ordering table below: RW16 Basic Stirrer (RW16), Eurostar Power Basic (Basic), Eurostar Power Control-Visc (Control-Visc), Eurostar Power Control-Visc P4 (P4), Eurostar Power Control-Visc P7 (P7), Eurostar Power Control-Visc 6000 (CV6000)
† PTFE-coated