Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Acclaim™ 5µm PepMap™ 300 µ-Precolumns™ Cartridge Columns

Thermo Scientific™ Acclaim™ 5µm PepMap™ 300 µ-Precolumns™ Cartridge Columns are very short microcolumns consisting of a set of disposable cartridges. Cartridge holder sold separately (p/n 164650).

$475.50 - $519.50

Specifications

For Use With LC/MS, ESI-MS, MALDI-MS
Stationary Phase C18
Column Type Peptide Analysis
Particle Size 5 µm
Pore Size 300 Å
View More Specs

Products 2
Catalog Number Mfr. No. Particle Size Diameter (Metric) Length (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Particle Size Diameter (Metric) Length (Metric) Price Quantity & Availability  
DX163592
View Documents
Thermo Scientific™
163592
5 µm 1 mm 5 mm Pack of 5 for $475.50
Add to cart
 
DX163593
View Documents
Thermo Scientific™
163593
5 µm 1 mm 15 mm Pack of 5 for $519.50
Add to cart
 
Description

Description

Thermo Scientific™ Acclaim™ 5µm PepMap™ 300 µ-Precolumns™ Cartridge Columns are very short microcolumns consisting of a set of disposable cartridges. Cartridge holder sold separately (p/n 164650).

  • For sample pre-concentration, sample cleanup, or in various column-switching techniques
  • 300Å pore size
Specifications

Specifications

LC/MS, ESI-MS, MALDI-MS
Peptide Analysis
300 Å
C18
5 µm
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit