Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ACCN1 Antibody, DyLight 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB1002531G
Description
ACCN1 Polyclonal antibody specifically detects ACCN1 in Human, Rat samples. It is validated for ImmunohistochemistrySpecifications
ACCN1 | |
Polyclonal | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Rat | |
Purified |
Immunohistochemistry | |
DyLight 488 | |
Acid-sensing ion channel 2, Amiloride-sensitive brain sodium channel, amiloride-sensitive cation channel 1, neuronal, ASIC2a, ASIC2Mammalian degenerin homolog, BNAC1, BNaC1ACCN, BNC1Amiloride-sensitive cation channel neuronal 1, degenerin, hBNaC1, MDEGBrain sodium channel 1, neuronal amiloride-sensitive cation channel 1 | |
CSGPGRFMAREQ (amino acid residues 15-26). | |
0.1 mL | |
Apoptosis, Cell Biology, Neuroscience | |
40 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction