Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ASIC5 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548928

Catalog No. PA548928

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Cation channel that gives rise to very low constitutive currents in the absence of activation. The activated channel exhibits selectivity for sodium, and is inhibited by amiloride.
Specifications

Specifications

ASIC5
Polyclonal
PBS with 0.09% sodium azide
Q9NY37
Acid-sensing ion channel 5, ASIC5, Amiloride-sensitive cation channel 5, Human intestine Na(+) channel, HINaC, ASIC5, ACCN5, HINAC
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
51802
Western Blot
Unconjugated
ASIC5
Liquid
ASIC5
KLH conjugated synthetic peptide between 388-414 amino acids from the C-terminal region of human ACCN5.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit